Merck
Correlation between the tumoral expression of beta3-integrin and outcome in cervical cancer patients who had undergone radiotherapy.
British journal of cancer (2004-12-15)
G Gruber, J Hess, C Stiefel, D M Aebersold, Y Zimmer, R H Greiner, U Studer, H J Altermatt, R Hlushchuk, V Djonov
ABSTRAKT
Integrins are cell-surface receptors, which mediate cell-to-cell and cell-to-extracellular matrix adhesion. Besides playing an important role in tumour angiogenesis, beta3-integrin is also expressed in several types of epithelial cancer cells. It was the purpose of the present study to evaluate the prognostic value of beta3-integrin expression in patients with cervical cancer. Biopsies were taken from 82 patients with squamous cell or adenocarcinomas of the uterine cervix who had undergone external-beam radiotherapy with or without brachytherapy. These tissue samples were analysed immunohistochemically for the expression of beta3-integrin. The impact of immunoreactivity for beta3-integrin on survival end points was assessed by univariate and multivariate analyses, and its correlation with clinicopathological characteristics evaluated by crosstabulations. beta3-integrin was expressed in 61% (50 of 82) of the patients. Kaplan-Meier curves revealed local progression-free survival, distant metastasis-free survival and cause-specific survival to be significantly shorter (P-values according to the log-rank test: 0.002, 0.04 and 0.01, respectively) in patients with beta3-integrin expression. The prognostic impact of this parameter was even higher than for other well-known prognostic parameters and remained statistically significant in the multivariate analyses. beta3-integrin, which is expressed in the majority of patients with advanced cervical cancer, has a significant prognostic impact on outcome according to univariate and multivariate analyses.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone SAP, clone SAP, Chemicon®, from mouse