Merck
Bacteriophage trigger antiviral immunity and prevent clearance of bacterial infection.
Science (New York, N.Y.) (2019-03-30)
Johanna M Sweere, Jonas D Van Belleghem, Heather Ishak, Michelle S Bach, Medeea Popescu, Vivekananda Sunkari, Gernot Kaber, Robert Manasherob, Gina A Suh, Xiou Cao, Christiaan R de Vries, Dung N Lam, Payton L Marshall, Maria Birukova, Ethan Katznelson, Daniel V Lazzareschi, Swathi Balaji, Sundeep G Keswani, Thomas R Hawn, Patrick R Secor, Paul L Bollyky
ABSTRAKT
Bacteriophage are abundant at sites of bacterial infection, but their effects on mammalian hosts are unclear. We have identified pathogenic roles for filamentous Pf bacteriophage produced by Pseudomonas aeruginosa (Pa) in suppression of immunity against bacterial infection. Pf promote Pa wound infection in mice and are associated with chronic human Pa wound infections. Murine and human leukocytes endocytose Pf, and internalization of this single-stranded DNA virus results in phage RNA production. This triggers Toll-like receptor 3 (TLR3)- and TIR domain-containing adapter-inducing interferon-β (TRIF)-dependent type I interferon production, inhibition of tumor necrosis factor (TNF), and the suppression of phagocytosis. Conversely, immunization of mice against Pf prevents Pa wound infection. Thus, Pf triggers maladaptive innate viral pattern-recognition responses, which impair bacterial clearance. Vaccination against phage virions represents a potential strategy to prevent bacterial infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)