Merck
Expression vector-derived heterogeneity in a therapeutic IgG4 monoclonal antibody.
mAbs (2018-10-27)
Douglas S Rehder, Chris J Wisniewski, Denfeng Liu, Diya Ren, Dell Farnan, Matthew R Schenauer
ABSTRAKT
While characterizing a therapeutic IgG4 monoclonal antibody (mAb), we observed a variant with a mass 1177 Da larger than the predominant mAb form that could not be ascribed to previously described modifications. Through successive rounds of experimentation, we localized the mass addition to the C-terminus of the heavy chain (HC). During this process we observed that when the mAb was broken down into separate domains, the Fc and the 1177 Da-modified Fc could be chromatographically separated. Separation allowed collection of native and modified Fc fractions for LC/MS peptide mapping. A unique peptide present in the modified fraction was de novo sequenced and demonstrated to be a modified form of the HC C-terminus lacking two native residues (GK) and gaining twelve additional non-native residues (EAEAASASELFQ). Aware of other mAb variants with genetic origins, we sought to understand whether this modification too had a genetic basis. In silico translation of the expression vector encoding the mAb demonstrated that a normally non-coding section of nucleotides in the + 1 reading frame relative to the HC C-terminal coding region could have led to a transcript with the non-native C-terminal extension. Two potential mechanisms for how this nucleotide sequence might have fused to the native HC coding region and led to expression of the extension product are presented.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%