Merck
The F-Box Domain-Dependent Activity of EMI1 Regulates PARPi Sensitivity in Triple-Negative Breast Cancers.
Molecular cell (2018-12-18)
Antonio Marzio, Joseph Puccini, Youngho Kwon, Natalia K Maverakis, Arnaldo Arbini, Patrick Sung, Dafna Bar-Sagi, Michele Pagano
ABSTRAKT
The BRCA1-BRCA2-RAD51 axis is essential for homologous recombination repair (HRR) and is frequently disrupted in breast cancers. PARP inhibitors (PARPis) are used clinically to treat BRCA-mutated breast tumors. Using a genetic screen, we identified EMI1 as a modulator of PARPi sensitivity in triple-negative breast cancer (TNBC) cells. This function requires the F-box domain of EMI1, through which EMI1 assembles a canonical SCF ubiquitin ligase complex that constitutively targets RAD51 for degradation. In response to genotoxic stress, CHK1-mediated phosphorylation of RAD51 counteracts EMI1-dependent degradation by enhancing RAD51's affinity for BRCA2, leading to RAD51 accumulation. Inhibition of RAD51 degradation restores HRR in BRCA1-depleted cells. Human breast cancer samples display an inverse correlation between EMI1 and RAD51 protein levels. A subset of BRCA1-deficient TNBC cells develop resistance to PARPi by downregulating EMI1 and restoring RAD51-dependent HRR. Notably, reconstitution of EMI1 expression reestablishes PARPi sensitivity both in cellular systems and in an orthotopic mouse model.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Hydroxyurea, 98%, powder
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-BRCA2 (Ab-1) Mouse mAb (2B), liquid, clone 2B, Calbiochem®