Merck
  • Optimization of the method for analyzing endocytosis of fluorescently tagged molecules: Impact of incubation in the cell culture medium and cell surface wash with glycine-hydrochloric acid buffer.
Optimization of the method for analyzing endocytosis of fluorescently tagged molecules: Impact of incubation in the cell culture medium and cell surface wash with glycine-hydrochloric acid buffer.
Journal of controlled release : official journal of the Controlled Release Society (2019-08-24)
Noriyasu Kamei, Satoshi Yamamoto, Hiro Hashimoto, Megumi Nishii, Moe Miyaura, Kiho Tomada, Ikuhiko Nakase, Mariko Takeda-Morishita
ABSTRAKT
To obtain the therapeutic effect of biological medicines, such as proteins and nucleic acids, these medicines must achieve their intracellular target, such as the cytoplasm, and pass through biological membrane barriers. Endocytosis is an attractive route for the intracellular delivery of such drugs, and various endocytosis inhibitors have been used as tools to study the involvement of endocytosis in the cell internalization of delivery carriers. However, the specificity of these inhibitors has been insufficiently studied, and our preliminary tests could not detect the expected effect of the well-known endocytosis inhibitors. Therefore, the present study aimed to optimize the experimental conditions to precisely analyze cellular internalization via endocytosis. We first found that incubation of model molecules, such as transferrin (Tf) and cholera toxin subunit B (CTB), in cell culture medium (DMEM) could efficiently induce their internalization to HeLa cells compared to that in transport buffer (HBSS). Moreover, we clarified that cell surface wash with glycine-hydrochloric acid buffer before confocal microscopy and flow cytometry strengthened the intracellular fluorescence of Tf, CTB, and dextran tagged with fluorescent probes possibly via the neutralization of endosomal pH. Even under the optimized condition, however, the specificity of endocytosis inhibitors was disputable. The present study suggested the importance of the optimization of the study design with endocytosis inhibitors in analyzing cellular internalization.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride