Merck
Inflammasome components Asc and caspase-1 mediate biomaterial-induced inflammation and foreign body response.
Proceedings of the National Academy of Sciences of the United States of America (2011-11-24)
Ahsan F Malik, Rafaz Hoque, Xinshou Ouyang, Ayaz Ghani, Enping Hong, Khadija Khan, Laura Beth Moore, Gilbert Ng, Fay Munro, Richard A Flavell, Yan Shi, Themis R Kyriakides, Wajahat Z Mehal
ABSTRAKT
Implantation of biomaterials and devices into soft tissues leads to the development of the foreign body response (FBR), which can interfere with implant function and eventually lead to failure. The FBR consists of overlapping acute and persistent inflammatory phases coupled with collagenous encapsulation and currently there are no therapeutic options. Initiation of the FBR involves macrophage activation, proceeding to giant cell formation, fibroblast activation, and collagen matrix deposition. Despite the recognition of this sequence of events, the molecular pathways required for the FBR have not been elucidated. We have identified that the acute inflammatory response to biomaterials requires nucleotide-binding domain and leucine-rich repeat-containing 3 (Nlrp3), apoptosis-associated speck-like protein containing CARD (Asc), and caspase-1, as well as plasma membrane cholesterol, and Syk signaling. Full development of the FBR is dependent on Asc and caspase-1, but not Nlrp3. The common antiinflammatory drug aspirin can reduce inflammasome activation and significantly reduce the FBR. Taken together, these findings expand the role of the inflammasome from one of sensing damage associated molecular patterns (DAMPs) to sensing all particulate matter irrespective of size. In addition, implication of the inflammasome in biomaterial recognition identifies key pathways, which can be targeted to limit the FBR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipopolysaccharides (rough strains) from Salmonella enterica serotype minnesota Re 595 (Re mutant)
Sigma-Aldrich
Piceatannol, powder