Merck
2-Deoxy-d-Glucose Ameliorates PKD Progression.
Journal of the American Society of Nephrology : JASN (2015-11-05)
Marco Chiaravalli, Isaline Rowe, Valeria Mannella, Giacomo Quilici, Tamara Canu, Veronica Bianchi, Antonia Gurgone, Sofia Antunes, Patrizia D'Adamo, Antonio Esposito, Giovanna Musco, Alessandra Boletta
ABSTRAKT
Autosomal dominant polycystic kidney disease (ADPKD) is an important cause of ESRD for which there exists no approved therapy in the United States. Defective glucose metabolism has been identified as a feature of ADPKD, and inhibition of glycolysis using glucose analogs ameliorates aggressive PKD in preclinical models. Here, we investigated the effects of chronic treatment with low doses of the glucose analog 2-deoxy-d-glucose (2DG) on ADPKD progression in orthologous and slowly progressive murine models created by inducible inactivation of the Pkd1 gene postnatally. As previously reported, early inactivation (postnatal days 11 and 12) of Pkd1 resulted in PKD developing within weeks, whereas late inactivation (postnatal days 25-28) resulted in PKD developing in months. Irrespective of the timing of Pkd1 gene inactivation, cystic kidneys showed enhanced uptake of (13)C-glucose and conversion to (13)C-lactate. Administration of 2DG restored normal renal levels of the phosphorylated forms of AMP-activated protein kinase and its target acetyl-CoA carboxylase. Furthermore, 2DG greatly retarded disease progression in both model systems, reducing the increase in total kidney volume and cystic index and markedly reducing CD45-positive cell infiltration. Notably, chronic administration of low doses (100 mg/kg 5 days per week) of 2DG did not result in any obvious sign of toxicity as assessed by analysis of brain and heart histology as well as behavioral tests. Our data provide proof of principle support for the use of 2DG as a therapeutic strategy in ADPKD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Harris Hematoxylin Solution, Modified
Sigma-Aldrich
Przeciwciało przeciwko karboksylazie fosfoacetylo-CoA (Ser79), Upstate®, from rabbit
Sigma-Aldrich
Przeciwciało anty-winkulina, klon V284, clone V284, Upstate®, from mouse
Sigma-Aldrich
Tamoxifen, ≥99%