Merck
Long noncoding RNA MALAT1 suppresses breast cancer metastasis.
Nature genetics (2018-10-24)
Jongchan Kim, Hai-Long Piao, Beom-Jun Kim, Fan Yao, Zhenbo Han, Yumeng Wang, Zhenna Xiao, Ashley N Siverly, Sarah E Lawhon, Baochau N Ton, Hyemin Lee, Zhicheng Zhou, Boyi Gan, Shinichi Nakagawa, Matthew J Ellis, Han Liang, Mien-Chie Hung, M James You, Yutong Sun, Li Ma
ABSTRAKT
MALAT1 has previously been described as a metastasis-promoting long noncoding RNA (lncRNA). We show here, however, that targeted inactivation of the Malat1 gene in a transgenic mouse model of breast cancer, without altering the expression of its adjacent genes, promotes lung metastasis, and that this phenotype can be reversed by genetic add-back of Malat1. Similarly, knockout of MALAT1 in human breast cancer cells induces their metastatic ability, which is reversed by re-expression of Malat1. Conversely, overexpression of Malat1 suppresses breast cancer metastasis in transgenic, xenograft, and syngeneic models. Mechanistically, the MALAT1 lncRNA binds and inactivates the prometastatic transcription factor TEAD, preventing TEAD from associating with its co-activator YAP and target gene promoters. Moreover, MALAT1 levels inversely correlate with breast cancer progression and metastatic ability. These findings demonstrate that MALAT1 is a metastasis-suppressing lncRNA rather than a metastasis promoter in breast cancer, calling for rectification of the model for this highly abundant and conserved lncRNA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2