Merck
Genistein protects hematopoietic stem cells against G-CSF-induced DNA damage.
Cancer prevention research (Philadelphia, Pa.) (2014-03-13)
Liliana R Souza, Erica Silva, Elissa Calloway, Omer Kucuk, Michael Rossi, Morgan L McLemore
ABSTRAKT
Granulocyte colony-stimulating factor (G-CSF) has been used to treat neutropenia in various clinical settings. Although clearly beneficial, there are concerns that the chronic use of G-CSF in certain conditions increases the risk of myelodysplastic syndrome (MDS) and/or acute myeloid leukemia (AML). The most striking example is in severe congenital neutropenia (SCN). Patients with SCN develop MDS/AML at a high rate that is directly correlated to the cumulative lifetime dosage of G-CSF. Myelodysplastic syndrome and AML that arise in these settings are commonly associated with chromosomal deletions. We have demonstrated in this study that chronic G-CSF treatment in mice results in expansion of the hematopoietic stem cell (HSC) population. In addition, primitive hematopoietic progenitors from G-CSF-treated mice show evidence of DNA damage as demonstrated by an increase in double-strand breaks and recurrent chromosomal deletions. Concurrent treatment with genistein, a natural soy isoflavone, limits DNA damage in this population. The protective effect of genistein seems to be related to its preferential inhibition of G-CSF-induced proliferation of HSCs. Importantly, genistein does not impair G-CSF-induced proliferation of committed hematopoietic progenitors, nor diminishes neutrophil production. The protective effect of genistein was accomplished with plasma levels that are attainable through dietary supplementation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo-H2A.X (Ser139), Upstate®, from rabbit