Merck
α-Synuclein strains target distinct brain regions and cell types.
Nature neuroscience (2019-12-04)
Angus Lau, Raphaella W L So, Heather H C Lau, Jason C Sang, Alejandro Ruiz-Riquelme, Shelaine C Fleck, Erica Stuart, Sindhu Menon, Naomi P Visanji, Georg Meisl, Rania Faidi, Maria M Marano, Cian Schmitt-Ulms, Zhilan Wang, Paul E Fraser, Anurag Tandon, Bradley T Hyman, Holger Wille, Martin Ingelsson, David Klenerman, Joel C Watts
ABSTRAKT
The clinical and pathological differences between synucleinopathies such as Parkinson's disease and multiple system atrophy have been postulated to stem from unique strains of α-synuclein aggregates, akin to what occurs in prion diseases. Here we demonstrate that inoculation of transgenic mice with different strains of recombinant or brain-derived α-synuclein aggregates produces clinically and pathologically distinct diseases. Strain-specific differences were observed in the signs of neurological illness, time to disease onset, morphology of cerebral α-synuclein deposits and the conformational properties of the induced aggregates. Moreover, different strains targeted distinct cellular populations and cell types within the brain, recapitulating the selective targeting observed among human synucleinopathies. Strain-specific clinical, pathological and biochemical differences were faithfully maintained after serial passaging, which implies that α-synuclein propagates via prion-like conformational templating. Thus, pathogenic α-synuclein exhibits key hallmarks of prion strains, which provides evidence that disease heterogeneity among the synucleinopathies is caused by distinct α-synuclein strains.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, powder, BioReagent, 30-350 units/mg protein (E1%/280), suitable for cell culture
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
DAPI, for nucleic acid staining