Merck
Treacle controls the nucleolar response to rDNA breaks via TOPBP1 recruitment and ATR activation.
Nature communications (2020-01-09)
Clémence Mooser, Ioanna-Eleni Symeonidou, Pia-Amata Leimbacher, Alison Ribeiro, Ann-Marie K Shorrocks, Stephanie Jungmichel, Sara C Larsen, Katja Knechtle, Arti Jasrotia, Diana Zurbriggen, Alain Jeanrenaud, Colin Leikauf, Daniel Fink, Michael L Nielsen, Andrew N Blackford, Manuel Stucki
ABSTRAKT
Induction of DNA double-strand breaks (DSBs) in ribosomal DNA (rDNA) repeats is associated with ATM-dependent repression of ribosomal RNA synthesis and large-scale reorganization of nucleolar architecture, but the signaling events that regulate these responses are largely elusive. Here we show that the nucleolar response to rDNA breaks is dependent on both ATM and ATR activity. We further demonstrate that ATM- and NBS1-dependent recruitment of TOPBP1 in the nucleoli is required for inhibition of ribosomal RNA synthesis and nucleolar segregation in response to rDNA breaks. Mechanistically, TOPBP1 recruitment is mediated by phosphorylation-dependent interactions between three of its BRCT domains and conserved phosphorylated Ser/Thr residues at the C-terminus of the nucleolar phosphoprotein Treacle. Our data thus reveal an important cooperation between TOPBP1 and Treacle in the signaling cascade that triggers transcriptional inhibition and nucleolar segregation in response to rDNA breaks.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-TCOF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Roche
Anty-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H2A.X (Ser139), klon JBW301, clone JBW301, Upstate®, from mouse