Merck
Reverse poly(ε-caprolactone)-g-dextran graft copolymers. Nano-carriers for intracellular uptake of anticancer drugs.
Carbohydrate polymers (2020-01-19)
Victor Delorme, Laure Lichon, Hana Mahindad, Sylvie Hunger, Nabila Laroui, Morgane Daurat, Anastasia Godefroy, Jean Coudane, Magali Gary-Bobo, Hélène Van Den Berghe
ABSTRAKT
A new fully biodegradable "reverse" oligosaccharide-based amphiphilic graft copolymer structure with a hydrophobic backbone and hydrophilic side chains, poly(ε-caprolactone)-g-dextran (PCL-g-Dex) was synthetized. For this purpose, "clickable" propargylated PCL (PCL-yne) and azido-dextran (Dex-N3) were prepared to further synthesize PCL-g-Dex copolymer by a Huisgen's cycloaddition. This "reverse" copolymer architecture self-assembled in biodegradable nano-carriers, in the shape of dynamic polymeric micelles, and were loaded with doxorubicin (Dox) anti-cancer drug. Dox-loaded micelles showed different drug releases depending on the pH. Cytotoxicity tests showed that Dox-loaded micelles can selectively kill colon cancer cells (HCT-116) while they have no cytotoxic effect towards healthy cells (CCD-45SK). Fluorescent micelles based on FITC-labelled PCL-g-Dex copolymer were used for fluorescence imaging and flow cytometry assays. These experiments proved the effective and specific internalization of micelles by cancer cells, whereas healthy cells showed a very poor uptake. These results show that PCL-g-Dex micelles may be a promising Dox nano-carrier in cancer chemotherapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%