Merck
The first potent and selective inhibitors of the glycine transporter type 2.
Journal of medicinal chemistry (2001-08-10)
W L Caulfield, I T Collie, R S Dickins, O Epemolu, R McGuire, D R Hill, G McVey, J R Morphy, Z Rankovic, H Sundaram
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ORG 25543 Hydrochloride, ≥95% (HPLC)