Merck
Enzymatic production of 4-O-methyl d-glucaric acid from hardwood xylan.
Biotechnology for biofuels (2020-03-20)
Thu V Vuong, Emma R Master
ABSTRAKT
Dicarboxylic acids offer several applications in detergent builder and biopolymer fields. One of these acids, 4-O-methyl d-glucaric acid, could potentially be produced from glucuronoxylans, which are a comparatively underused fraction of wood and agricultural biorefineries. Accordingly, an enzymatic pathway was developed that combines AxyAgu115A, a GH115 α-glucuronidase from Amphibacillus xylanus, and GOOX, an AA7 gluco-oligosaccharide oxidase from Sarocladium strictum, to produce this bio-based chemical from glucuronoxylan. AxyAgu115A was able to release almost all 4-O-methyl d-glucuronic acid from glucuronoxylan while a GOOX variant, GOOX-Y300A, could convert 4-O-methyl d-glucuronic acid to the corresponding glucaric acid at a yield of 62%. Both enzymes worked effectively at alkaline conditions that increase xylan solubility. Given the sensitivity of AxyAgu115A to hydrogen peroxide and optimal performance of GOOX-Y300A at substrate concentrations above 20 mM, the two-step enzyme pathway was demonstrated as a sequential, one-pot reaction. Additionally, the resulting xylan was easily recovered from the one-pot reaction, and it was enzymatically hydrolysable. The pathway in this study requires only two enzymes while avoiding a supplementation of costly cofactors. This cell-free approach provides a new strategy to make use of the underutilized hemicellulose stream from wood and agricultural biorefineries.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)