Merck
Molecular alterations of the TLR4-signaling cascade in canine epilepsy.
BMC veterinary research (2020-01-22)
Eva-Lotta von Rüden, Fabio Gualtieri, Katharina Schönhoff, Maria Reiber, Fabio Wolf, Wolfgang Baumgärtner, Florian Hansmann, Andrea Tipold, Heidrun Potschka
ABSTRAKT
Cumulating evidence from rodent models points to a pathophysiological role of inflammatory signaling in the epileptic brain with Toll-like receptor-4 signaling acting as one key factor. However, there is an apparent lack of information about expression alterations affecting this pathway in canine patients with epilepsy. Therefore, we have analyzed the expression pattern of Toll-like receptor 4 and its ligands in brain tissue of canine patients with structural or idiopathic epilepsy in comparison with tissue from laboratory dogs or from owner-kept dogs without neurological diseases. The analysis revealed an overexpression of Toll-like receptor-4 in the CA3 region of dogs with structural epilepsy. Further analysis provided evidence for an upregulation of Toll-like receptor-4 ligands with high mobility group box-1 exhibiting increased expression levels in the CA1 region of dogs with idiopathic and structural epilepsy, and heat shock protein 70 exhibiting increased expression levels in the piriform lobe of dogs with idiopathic epilepsy. In further brain regions, receptor and ligand expression rates proved to be either in the control range or reduced below control levels. Our study reveals complex molecular alterations affecting the Toll-like receptor signaling cascade, which differ between epilepsy types and between brain regions. Taken together, the data indicate that multi-targeting approaches modulating Toll-like receptor-4 signaling might be of interest for management of canine epilepsy. Further studies are recommended to explore respective molecular alterations in more detail in dogs with different etiologies and to confirm the role of the pro-inflammatory signaling cascade as a putative target.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 15 mL
Sigma-Aldrich
TWEEN® 20, Molecular Biology, viscous liquid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse