Merck
  • Cellulose ether treatment in vivo generates chronic wasting disease prions with reduced protease resistance and delayed disease progression.
Cellulose ether treatment in vivo generates chronic wasting disease prions with reduced protease resistance and delayed disease progression.
Journal of neurochemistry (2019-09-26)
Samia Hannaoui, Maria Immaculata Arifin, Sheng Chun Chang, Jie Yu, Preetha Gopalakrishnan, Katsumi Doh-Ura, Hermann M Schatzl, Sabine Gilch
ABSTRAKT
Chronic wasting disease (CWD) is a prion disease of free-ranging and farmed cervids that is highly contagious because of extensive prion shedding and prion persistence in the environment. Previously, cellulose ether compounds (CEs) have been shown to significantly extend the survival of mice inoculated with mouse-adapted prion strains. In this study, we used CEs, TC-5RW, and 60SH-50, in vitro and in vivo to assess their efficacy to interfere with CWD prion propagation. In vitro, CEs inhibited CWD prion amplification in a dose-dependent manner. Transgenic mice over-expressing elk PrPC (tgElk) were injected subcutaneously with a single dose of either of the CEs, followed by intracerebral inoculation with different CWD isolates from white tailed deer, mule deer, or elk. All treated groups showed a prolonged survival of up to more than 30 % when compared to the control group regardless of the CWD isolate used for infection. The extended survival in the treated groups correlated with reduced proteinase K resistance of prions. Remarkably, passage of brain homogenates from treated or untreated animals in tgElk mice resulted in a prolonged life span of mice inoculated with homogenates from CE-treated mice (of + 17%) even in the absence of further treatment. Besides the delayed disease onset upon passage in TgElk mice, the reduced proteinase K resistance was maintained but less pronounced. Therefore, these compounds can be very useful in limiting the spread of CWD in captive and wild-ranging cervids.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Humic acid, technical
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Sodium phosphate, 96%