Merck
  • Human umbilical cord mesenchymal stem cell-derived and dermal fibroblast-derived extracellular vesicles protect dermal fibroblasts from ultraviolet radiation-induced photoaging in vitro.
Human umbilical cord mesenchymal stem cell-derived and dermal fibroblast-derived extracellular vesicles protect dermal fibroblasts from ultraviolet radiation-induced photoaging in vitro.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2020-03-04)
Mingwu Deng, Ziyou Yu, Dong Li, Xiangsheng Wang, Guangdong Zhou, Wei Liu, Yilin Cao, Wanyao Xia, Wei Li, Wen Jie Zhang
ABSTRAKT
Ultraviolet B (UVB) radiation is a major cause of aging in dermal fibroblasts. Human umbilical cord mesenchymal stem cell-derived extracellular vesicles (MSC-EVs) show antioxidant activity. In this study, the anti-aging effects of MSC-EVs on dermal fibroblast photoaging induced by UVB radiation were evaluated, and the effects of extracellular vesicles derived from dermal fibroblasts (Fb-EVs) were compared. Human umbilical cord mesenchymal stem cells and human dermal fibroblasts were cultured, and MSC-EVs and Fb-EVs were isolated and characterized. Human dermal fibroblasts were cultured in the absence or presence of different concentrations of EVs 24 hours prior to UVB radiation exposure. Cell proliferation and cell cycle were evaluated, and senescent cells and intracellular ROS were detected. The expressions of matrix metalloproteinase-1 (MMP-1), extracellular matrix protein collagen type 1 (Col-1), and antioxidant proteins such as glutathione peroxidase 1 (GPX-1), superoxide dismutase (SOD), and catalase were also analyzed. Pretreatment with MSC-EVs or Fb-EVs significantly inhibited the production of ROS induced by UVB radiation, increased dermal fibroblast proliferation, protected cells against UVB-induced cell death and cell cycle arrest, and remarkably decreased the percentage of aged cells. Pretreatment with MSC-EVs or Fb-EVs promoted the expressions of GPX-1 and Col-1 and decreased the expression of MMP-1. Both MSC-EVs and Fb-EVs protected dermal fibroblasts from UVB-induced photoaging, likely through their antioxidant activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder