Merck
Apelin Promotes ECM Synthesis by Enhancing Autophagy Flux via TFEB in Human Degenerative NP Cells under Oxidative Stress.
BioMed research international (2020-03-10)
Wei Jiang, Pei Zhao, Xuemei Zhang
ABSTRAKT
Apelin alleviates oxidative stress which contributes to the development of aging. IVDD is a disease closely correlated to aging and oxidative stress which is known to be harmful to NP cells' matrix synthesis. The purpose of the present study was to investigate the role and underlying mechanism of Apelin in NP cells' matrix degradation under oxidative stress. First, the mRNA and protein expressions of Apelin were checked by RT-PCR and Western blot in NP from normal and degenerative IVD to explore the relationship between Apelin and IVDD preliminarily. Then, H2O2 was used to mimic oxidative stress of NP cells. After treated with Apelin 13 and CQ, the GAG content was assessed by DMMB and the mRNA/protein expressions of NP matrix macromolecules (Collagen II and Aggrecan) and autophagy-related markers (LC3 and p62) were assessed by RT-PCR/Western blot. Finally, TFEB was knocked down by esiRNA-TFEB transfection and the nucleoprotein expression of TFEB and autophagy-related markers (LC3 and p62) were assessed by Western blot to discuss whether TFEB is involved in Apelin regulating autophagy flux in NP cells under oxidative stress. Our data first confirmed that the mRNA and protein expressions of Apelin were decreased with IVDD. Furthermore, Apelin increased GAG content of NP cells and mRNA/protein expressions of NP matrix macromolecules (Collagen II and Aggrecan) and promoted autophagic flux (LC3II/I increased and p62 decreased) under oxidative stress. Finally, after transfected with esiRNA-TFEB, Apelin cannot promote autophagic flux any more in human degenerative NP cells. Our data indicated that Apelin promotes ECM synthesis by enhancing autophagy flux via TFEB in human degenerative NP cells under oxidative stress. This viewpoint may provide a new therapeutic idea for IVDD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %
Sigma-Aldrich
MISSION® esiRNA, targeting human TFEB