Merck
Tumor suppressor death-associated protein kinase 1 inhibits necroptosis by p38 MAPK activation.
Cell death & disease (2020-05-06)
Yung-Hsuan Wu, Ting-Fang Chou, Leslie Young, Fu-Yi Hsieh, Hsuan-Yin Pan, Shu-Ting Mo, Shani Bialik Brown, Ruey-Hwa Chen, Adi Kimchi, Ming-Zong Lai
ABSTRAKT
Death-associated protein kinase 1 (DAPK1, DAPk, DAPK) is known for its involvement in apoptosis and autophagy-associated cell death. Here, we identified an unexpected function of DAPK1 in suppressing necroptosis. DAPK1-deficiency renders macrophages and dendritic cells susceptible to necroptotic death. We also observed an inhibitory role for DAPK1 in necroptosis in HT-29 cells, since knockdown or knockout of DAPK1 in such cells increased their sensitivity to necroptosis. Increased necroptosis was associated with enhanced formation of the RIPK1-RIPK3-MLKL complex in these DAPK1-deficient cells. We further found that DAPK1-deficiency led to decreased MAPK activated kinase 2 (MK2) activation and reduced RIPK1 S321 phosphorylation, with this latter representing a critical step controlling necrosome formation. Most TNF signaling pathways, including ERK, JNK, and AKT, were not regulated by DAPK. In contrast, DAPK bound p38 MAPK and selectively promoted p38 MAPK activation, resulting in enhanced MK2 phosphorylation. Our results reveal a novel role for DAPK1 in inhibiting necroptosis and illustrate an unexpected selectivity for DAPK1 in promoting p38 MAPK-MK2 activation. Importantly, our study suggests that modulation of necroptosis and p38/MK2-mediated inflammation may be achieved by targeting DAPK1.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-MLKL Antibody, clone 3H1, clone 3H1, from rat
Sigma-Aldrich
Przeciwciało anty-FADD, klon 1F7, clone 1F7, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Death Associated Protein Kinase antibody produced in mouse, clone DAPK-55, ascites fluid
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)