Merck
  • Microtubule disrupting agent-mediated inhibition of cancer cell growth is associated with blockade of autophagic flux and simultaneous induction of apoptosis.
Microtubule disrupting agent-mediated inhibition of cancer cell growth is associated with blockade of autophagic flux and simultaneous induction of apoptosis.
Cell proliferation (2020-03-14)
Mohammad Hasanain, Rohit Sahai, Praveen Pandey, Mayank Maheshwari, Kuldeep Choyal, Deepa Gandhi, Akhilesh Singh, Kavita Singh, Kalyan Mitra, Dipak Datta, Jayanta Sarkar
ABSTRAKT
Given that autophagy inhibition is a feasible way to enhance sensitivity of cancer cells towards chemotherapeutic agents, identifying potent autophagy inhibitor has obvious clinical relevance. Here, we investigated ability of TN-16, a microtubule disrupting agent, on modulation of autophagic flux and its significance in promoting in vitro and in vivo cancer cell death. The effect of TN-16 on cancer cell proliferation, cell division, autophagic process and apoptotic signalling was assessed by various biochemical (Western blot and SRB assay), morphological (TEM, SEM, confocal microscopy) and flowcytometric assays. In vivo anti-tumour efficacy of TN-16 was investigated in syngeneic mouse model of breast cancer. TN-16 inhibited cancer cell proliferation by impairing late-stage autophagy and induction of apoptosis. Inhibition of autophagic flux was demonstrated by accumulation of autophagy-specific substrate p62 and lack of additional LC3-II turnover in the presence of lysosomotropic agent. The effect was validated by confocal micrographs showing diminished autophagosome-lysosome fusion. Further studies revealed that TN-16-mediated inhibition of autophagic flux promotes apoptotic cell death. Consistent with in vitro data, results of our in vivo study revealed that TN-16-mediated tumour growth suppression is associated with blockade of autophagic flux and enhanced apoptosis. Our data signify that TN-16 is a potent autophagy flux inhibitor and might be suitable for (pre-) clinical use as standard inhibitor of autophagy with anti-cancer activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
TN-16, ≥98% (HPLC)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)