Merck
Stimulation of cortical bone formation with thienopyrimidine based inhibitors of Notum Pectinacetylesterase.
Bioorganic & medicinal chemistry letters (2016-02-22)
James E Tarver, Praveen K Pabba, Joseph Barbosa, Qiang Han, Michael W Gardyan, Robert Brommage, Andrea Y Thompson, James M Schmidt, Alan G E Wilson, Wei He, Victoria K Lombardo, Kenneth G Carson
ABSTRAKT
A group of small molecule thienopyrimidine inhibitors of Notum Pectinacetylesterase are described. We explored both 2-((5,6-thieno[2,3-d]pyrimidin-4-yl)thio)acetic acids and 2-((6,7-thieno[3,2-d]pyrimidin-4-yl)thio)acetic acids. In both series, highly potent, orally active Notum Pectinacetylesterase inhibitors were identified.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
LP-922056, ≥98% (HPLC)