- Tissue-specific tumor suppressor activity of retinoblastoma gene homologs p107 and p130.
Tissue-specific tumor suppressor activity of retinoblastoma gene homologs p107 and p130.
Genes & development (2004-12-03)
Jan-Hermen Dannenberg, Leontine Schuijff, Marleen Dekker, Martin van der Valk, Hein te Riele
PMID15574596
ABSTRAKT
The retinoblastoma gene family consists of three genes: RB, p107, and p130. While loss of pRB causes retinoblastoma in humans and pituitary gland tumors in mice, tumorigenesis in other tissues may be suppressed by p107 and p130. To test this hypothesis, we have generated chimeric mice from embryonic stem cells carrying compound loss-of-function mutations in the Rb gene family. We found that Rb/p107- and Rb/p130-deficient mice were highly cancer prone. We conclude that in a variety of tissues tumor development by loss of pRB is suppressed by its homologs p107 and p130. The redundancy of the retinoblastoma proteins in vivo is reflected by the behavior of Rb-family-defective mouse embryonic fibroblasts in vitro.