Merck
Nonsense-mediated mRNA decay factor UPF1 promotes aggresome formation.
Nature communications (2020-06-21)
Yeonkyoung Park, Joori Park, Hyun Jung Hwang, Byungju Kim, Kwon Jeong, Jeeyoon Chang, Jong-Bong Lee, Yoon Ki Kim
ABSTRAKT
Nonsense-mediated mRNA decay (NMD) typifies an mRNA surveillance pathway. Because NMD necessitates a translation event to recognize a premature termination codon on mRNAs, truncated misfolded polypeptides (NMD-polypeptides) could potentially be generated from NMD substrates as byproducts. Here, we show that when the ubiquitin-proteasome system is overwhelmed, various misfolded polypeptides including NMD-polypeptides accumulate in the aggresome: a perinuclear nonmembranous compartment eventually cleared by autophagy. Hyperphosphorylation of the key NMD factor UPF1 is required for selective targeting of the misfolded polypeptide aggregates toward the aggresome via the CTIF-eEF1A1-DCTN1 complex: the aggresome-targeting cellular machinery. Visualization at a single-particle level reveals that UPF1 increases the frequency and fidelity of movement of CTIF aggregates toward the aggresome. Furthermore, the apoptosis induced by proteotoxic stresses is suppressed by UPF1 hyperphosphorylation. Altogether, our data provide evidence that UPF1 functions in the regulation of a protein surveillance as well as an mRNA quality control.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Mysz anty-c-Myc (Ab-1) mAb (9E10), lyophilized, clone 9E10, Calbiochem®
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Benzamidine hydrochloride, 99%
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Przeciwciało anty-EF1α, klon CBP-KK1, clone CBP-KK1, Upstate®, from mouse