Merck
Purification and biochemical characterization of a novel secretory dipeptidyl peptidase IV from porcine serum.
Molecular and cellular biochemistry (2020-06-25)
Divya Kumar, Vivek K Hamse, K N Neema, Priya Babu Shubha, D M Chetan, Nanjunda Swamy Shivananju
ABSTRAKT
Purification of DPP-IV enzyme from porcine serum, is presented in this study for the first time. The high molecular weight DPP-IV from porcine serum was fractioned using Sephadex G-75 gel filtration followed by DEAE Sephadex anion exchange and Sephadex G-100 gel filtration chromatography columns with a final yield of 11.25%. The SDS-PAGE of the purified sample showed a single band of molecular mass nearing 160 kDa. Distinct single band was observed after PAS staining confirmed it to be a glycoprotein. The purified enzyme showed an optimum pH and temperature of 8 and 37 °C, respectively. The enzyme effectively cleaved fluorogenic substrate Gly-Pro-AMC with Km and Vmax of 4.578 µM and 90.84 nmoles/min, respectively. Purified DPP-IV activity was inhibited by Diprotin A with an IC50 value of 8.473 µM. Among the three plant extracts used to study DPP-IV inhibition, the aqueous hot extract of Terminalia chebula showed the highest inhibition of 87.19%, followed by the aqueous cold extract of Momordica carantia, ( 31.6%) and Azadirachta indica (34.16%) at the concentration of 25 µg.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ile-Pro-Ile, ≥97% (HPLC)