Merck
Pharmacological and genetic characterisation of the canine P2X4 receptor.
British journal of pharmacology (2020-02-06)
Reece A Sophocleous, Tracey Berg, Rocio K Finol-Urdaneta, Vanessa Sluyter, Shikara Keshiya, Lachlan Bell, Stephen J Curtis, Belinda L Curtis, Aine Seavers, Rachael Bartlett, Mark Dowton, Leanne Stokes, Lezanne Ooi, Ronald Sluyter
ABSTRAKT
P2X4 receptors are emerging therapeutic targets for treating chronic pain and cardiovascular disease. Dogs are well-recognised natural models of human disease, but information regarding P2X4 receptors in dogs is lacking. To aid the development and validation of P2X4 receptor ligands, we have characterised and compared canine and human P2X4 receptors. Genomic DNA was extracted from whole blood samples from 101 randomly selected dogs and sequenced across the P2RX4 gene to identify potential missense variants. Recombinant canine and human P2X4 receptors tagged with Emerald GFP were expressed in 1321N1 and HEK293 cells and analysed by immunoblotting and confocal microscopy. In these cells, receptor pharmacology was characterised using nucleotide-induced Fura-2 AM measurements of intracellular Ca2+ and known P2X4 receptor antagonists. P2X4 receptor-mediated inward currents in HEK293 cells were assessed by automated patch clamp. No P2RX4 missense variants were identified in any canine samples. Canine and human P2X4 receptors were localised primarily to lysosomal compartments. ATP was the primary agonist of canine P2X4 receptors with near identical efficacy and potency at human receptors. 2'(3')-O-(4-benzoylbenzoyl)-ATP, but not ADP, was a partial agonist with reduced potency for canine P2X4 receptors compared to the human orthologues. Five antagonists inhibited canine P2X4 receptors, with 1-(2,6-dibromo-4-isopropyl-phenyl)-3-(3-pyridyl)urea displaying reduced sensitivity and potency at canine P2X4 receptors. P2X4 receptors are highly conserved across dog pedigrees and display expression patterns and pharmacological profiles similar to human receptors, supporting validation and use of therapeutic agents for P2X4 receptor-related disease onset and management in dogs and humans.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
2′(3′)-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate triethylammonium salt, ≥93%
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Sigma-Aldrich
5-BDBD, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
(S)-Duloxetine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)