Merck
Prolactin-Responsive Circular RNA circHIPK3 Promotes Proliferation of Mammary Epithelial Cells from Dairy Cow.
Genes (2020-04-05)
Jia Liu, MoLan Zhang, DeWei Li, MengLu Li, LingHao Kong, MengWen Cao, YanHong Wang, ChengChuang Song, XingTang Fang, Hong Chen, Hu Xu, ChunLei Zhang
ABSTRAKT
The highly expressed circHIPK3 is a circular RNA that has been previously reported to regulate the growth of human cells. In this study, we found an increased expression of circHIPK3 in bovine mammary epithelial cells treated with prolactin (PRL) in high-throughput sequencing data. Thus, we further investigated the effect of circHIPK3 on the proliferation and differentiation of mammary epithelial cells. We used qRT-PCR/Cell Counting Kit-8 (CCK-8) and a Western blotting analysis to evaluate the effects on cell proliferation. We found that circHIPK3 promotes the proliferation of mammary epithelial cells. The STAT5 signaling pathway was previously associated with the prolactin response and when the STAT5 was suppressed, the expression of circHIPK3 decreased. The results suggest that the response to prolactin and the associated STAT5 signaling pathway affect the expression of circHIPK3, which subsequently affects the proliferation of mammary epithelial cells in dairy cows.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human HIPK3
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder