Merck
Sennoside A Induces GLP-1 Secretion Through Activation of the ERK1/2 Pathway in L-Cells.
Diabetes, metabolic syndrome and obesity : targets and therapy (2020-05-20)
Li Ma, Xinyu Cao, Xiaotong Ye, Jianping Ye, Yongning Sun
ABSTRAKT
Glucagon-like peptide-1 (GLP-1) is secreted from the intestinal L-cells to stimulate insulin secretion in the blood glucose control. Our previous study indicates that Sennoside A (SA) can increase the plasma GLP-1 level in a mouse model of type 2 diabetes. However, the mechanism of SA activity remains largely unknown. This issue was explored in this study. C57BL/6 mice were randomly divided into four groups: a control group without drug treatment, and the other groups with different SA dosages, respectively. Blood glucose was assayed by oral glucose tolerance test (OGTT). Plasma GLP-1 and insulin were investigated. Colon tissues were collected for mRNA or Western blot analysis. Immunofluorescence staining assays were performed to evaluate the number of β-cells and L-cells. In NCI-H716 cells, extracellular signal-regulated kinase 1/2 (ERK1/2) inhibitors were employed to investigate the SA-induced GLP-1 secretion mechanism. In this work, the SA was found to improve OGTT in mice. Plasma GLP-1 and insulin were markedly elevated by SA at the dosage of 45 mg/kg/day. Meanwhile, the increased phosphorylation status of EKR1/2 and prohormone convertase 1/3 (PC1/3) proteins were observed in the colon of SA-treated mice. The number of L-cells exhibited no change in each group. In the NCI-H716 cells, GLP-1 secretion induced by SA was blocked by the ERK1/2 inhibitor. The present study provides a direct evidence for the interaction between SA and L cells for induction of GLP-1 secretion. These data suggest that GLP-1 secretion induced by SA is dependent on the ERK1/2 signaling pathway. Therefore, the SA is a new drug candidate for the type 2 diabetes treatment by induction of GLP-1 secretion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ile-Pro-Ile, ≥97% (HPLC)
Sigma-Aldrich
Mouse KC / CXCL1 ELISA Kit, for serum, plasma and cell culture supernatant