Merck
Phagosomal removal of fungal melanin reprograms macrophage metabolism to promote antifungal immunity.
Nature communications (2020-05-10)
Samuel M Gonçalves, Cláudio Duarte-Oliveira, Cláudia F Campos, Vishukumar Aimanianda, Rob Ter Horst, Luis Leite, Toine Mercier, Paulo Pereira, Miguel Fernández-García, Daniela Antunes, Cláudia S Rodrigues, Catarina Barbosa-Matos, Joana Gaifem, Inês Mesquita, António Marques, Nuno S Osório, Egídio Torrado, Fernando Rodrigues, Sandra Costa, Leo Ab Joosten, Katrien Lagrou, Johan Maertens, João F Lacerda, António Campos, Gordon D Brown, Axel A Brakhage, Coral Barbas, Ricardo Silvestre, Frank L van de Veerdonk, Georgios Chamilos, Mihai G Netea, Jean-Paul Latgé, Cristina Cunha, Agostinho Carvalho
ABSTRAKT
In response to infection, macrophages adapt their metabolism rapidly to enhance glycolysis and fuel specialized antimicrobial effector functions. Here we show that fungal melanin is an essential molecule required for the metabolic rewiring of macrophages during infection with the fungal pathogen Aspergillus fumigatus. Using pharmacological and genetic tools, we reveal a molecular link between calcium sequestration by melanin inside the phagosome and induction of glycolysis required for efficient innate immune responses. By remodeling the intracellular calcium machinery and impairing signaling via calmodulin, melanin drives an immunometabolic signaling axis towards glycolysis with activation of hypoxia-inducible factor 1 subunit alpha (HIF-1α) and phagosomal recruitment of mammalian target of rapamycin (mTOR). These data demonstrate a pivotal mechanism in the immunometabolic regulation of macrophages during fungal infection and highlight the metabolic repurposing of immune cells as a potential therapeutic strategy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
MISSION® esiRNA, targeting human STIM1
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
Tributylamine, puriss. plus, ≥99.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film