Merck
  • Cabbage stem flea beetle's (Psylliodes chrysocephala L.) susceptibility to pyrethroids and tolerance to thiacloprid in the Czech Republic.
Cabbage stem flea beetle's (Psylliodes chrysocephala L.) susceptibility to pyrethroids and tolerance to thiacloprid in the Czech Republic.
PloS one (2019-09-21)
Jitka Stará, František Kocourek
ABSTRAKT
The cabbage stem flea beetle (CSFB), Psylliodes chrysocephala (Coleoptera: Chrysomelidae), has recently become a major pest species in winter oilseed rape in the Czech Republic. The susceptibility of CSFB populations from two localities to six pyrethroids, two neonicotinoids, one organophosphate and one oxadiazine was evaluated in 2015-2018 in glass vial experiments. The susceptibility of CSFB to thiacloprid and thiamethoxam was evaluated in feeding experiment in 2017 and 2018. High susceptibility of CSFB populations to lambda-cyhalothrin, cypermethrin, esfenvalerate, tau-fluvalinate, etofenprox, deltamethrin, chlorpyrifos, indoxacarb and acetamiprid was observed in the glass vial experiments. The LC50 and LC90 data obtained for pyrethroids in these experiments in 2015 represent baseline for CSFB resistance monitoring to pyrethroids in the Czech Republic. High tolerance of CSFB to thiacloprid of CSFB was demonstrated in glass vial and the feeding experiment, too.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Thiacloprid, PESTANAL®, analytical standard
Supelco
Indoxacarb, PESTANAL®, analytical standard
Supelco
Etofenprox, PESTANAL®, analytical standard
Supelco
Esfenvalerate, PESTANAL®, analytical standard
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard