Merck
2-Deoxy-D-glucose enhances the anti-cancer effects of idarubicin on idarubicin-resistant P388 leukemia cells.
Oncology letters (2020-06-23)
Taisuke Matsuo, Yumiko Konya, Eri Hirayama, Yasuyuki Sadzuka
ABSTRAKT
Cancer cells switch from mitochondrial oxidative phosphorylation to glycolysis, even in the presence of normal oxygen concentrations. Inhibition of the glycolytic pathway is therefore a critical strategy in cancer therapy. A non-metabolic glucose analog, 2-deoxy-D-glucose (2-DG), has been the focus of research on glycolytic inhibitors for use in cancer treatment. The current study examined the anti-cancer effects of 2-DG on idarubicin (IDA)-resistant P388 (P388/IDA) leukemia cells. P388/IDA cells were established following continuous exposure of IDA to P388 cells. Characterization of P388/IDA cells revealed increased lactate production and glucose consumption compared with P388 parent cells. The results of a cell viability assay determined that 2-DG induces higher toxicity in P388/IDA cells compared with P388 cells. Although 2-DG also exhibits endoplasmic reticulum (ER) stress-inducing activity, the cytotoxic effect of the ER stress inducer, tunicamycin, on P388/IDA cells was lower than that of P388 cells. A combination of 2-DG and IDA enhanced P388/IDA cell death compared with each agent alone. The results indicated that P388 cells activated glycolysis after acquiring IDA resistance and therefore, inhibition of the glycolytic pathway via 2-DG might be a useful strategy for cancer therapy against IDA- resistant leukemia cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
Tunicamycin from Streptomyces sp.