Merck
  • No association of complement mannose-binding lectin deficiency with cardiovascular disease in patients with Systemic Lupus Erythematosus.
No association of complement mannose-binding lectin deficiency with cardiovascular disease in patients with Systemic Lupus Erythematosus.
Scientific reports (2020-03-01)
A Kieninger-Gräfitsch, S Vogt, C Ribi, D Dubler, C Chizzolini, U Huynh-Do, M Osthoff, M Trendelenburg
ABSTRAKT
Cardiovascular (CV) morbidity is the major cause of death in patients with Systemic Lupus Erythematosus (SLE). Previous studies on mannose-binding lectin (MBL) gene polymorphisms in SLE patients suggest that low levels of complement MBL are associated with cardiovascular disease (CVD). However, as large studies on MBL deficiency based on resulting MBL plasma concentrations are lacking, the aim of our study was to analyze the association of MBL concentrations with CVD in SLE patients. Plasma MBL levels SLE patients included in the Swiss SLE Cohort Study were quantified by ELISA. Five different CV organ manifestations were documented. Of 373 included patients (85.5% female) 62 patients had at least one CV manifestation. Patients with MBL deficiency (levels below 500 ng/ml or 1000 ng/ml) had no significantly increased frequency of CVD (19.4% vs. 15.2%, P = 0.3 or 17.7% vs. 15.7%, P = 0.7). After adjustment for traditional CV risk factors, MBL levels and positive antiphospholipid serology (APL+) a significant association of CVD with age, hypertension, disease duration and APL+ was demonstrated. In our study of a large cohort of patients with SLE, we could not confirm previous studies suggesting MBL deficiency to be associated with an increased risk for CVD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
Mannan from Saccharomyces cerevisiae, prepared by alkaline extraction