Merck
  • Upregulation of Glutaredoxin 2 alleviates oxygen-glucose deprivation/reoxygenation-induced apoptosis and ROS production in neurons by enhancing Nrf2 signaling via modulation of GSK-3β.
Upregulation of Glutaredoxin 2 alleviates oxygen-glucose deprivation/reoxygenation-induced apoptosis and ROS production in neurons by enhancing Nrf2 signaling via modulation of GSK-3β.
Brain research (2020-06-12)
Jian Wen, Xin Li, Shaohua Zheng, Ying Xiao
ABSTRAKT
Glutaredoxin 2 (GRX2) is an antioxidative protein that exerts a key role in various pathological processes. However, whether GRX2 participates in modulating the oxidative stress during cerebral ischemia/reperfusion, injury is undermined. This study aimed to determine the potential role of GRX2 in regulating oxidative stress in cultured neurons induced by oxygen-glucose deprivation/reoxygenation (OGD/R), a cellular model for study of cerebral ischemia/reperfusion injury in vitro. Here, we showed that GRX2 expression was decreased in neurons subjected to OGD/R exposure. The upregulation of GRX2 markedly improved the viability of OGD/R-exposed neurons and caused a marked reduction in OGD/R-induced apoptosis and reactive oxygen species (ROS) production. On the contrary, depletion of GRX2 exacerbated the OGD/R-induced apoptosis and ROS production in cultured neurons. Moreover, GRX2 upregulation increased nuclear expression of nuclear factor erythroid 2-related factor 2 (Nrf2) and enhanced the activation of Nrf2/ARE signaling associated with modulation of glycogen synthase kinase-3β (GSK-3β) inhibition. Notably, inhibition of Nrf2 markedly abrogated GRX2-mediated protection against OGD/R-induced apoptosis and oxidative stress. Overall, these findings elucidate that GRX2 plays an essential role in regulating the protection against OGD/R-induced apoptosis and oxidative stress in neurons associated with its ability to enhance the activation of Nrf2 via modulation of GSK-3β. Our study indicates that GRX2 may play a key role in modulating neuronal apoptosis and oxidative stress induced by cerebral ischemia/reperfusion injury.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human GLRX2