Merck
  • Real-life multidisciplinary treatment for unresectable colorectal cancer liver metastases including hepatic artery infusion with chemo-filtration and liquid biopsy precision oncotherapy: observational cohort study.
Real-life multidisciplinary treatment for unresectable colorectal cancer liver metastases including hepatic artery infusion with chemo-filtration and liquid biopsy precision oncotherapy: observational cohort study.
Journal of cancer research and clinical oncology (2020-02-24)
Stefano Guadagni, Marco Clementi, Andrew R Mackay, Enrico Ricevuto, Giammaria Fiorentini, Donatella Sarti, Paola Palumbo, Panagiotis Apostolou, Ioannis Papasotiriou, Francesco Masedu, Marco Valenti, Aldo Victor Giordano, Gemma Bruera
ABSTRAKT
Hepatic artery infusion (HAI) and drug selection by liquid biopsy precision oncotherapy are under investigation for the multidisciplinary treatment of unresectable colorectal liver metastases (CRCLM) in progression after systemic therapy. Here, we compare the safety and efficacy of third-line HAI followed by target therapy with drug regimes selected by liquid biopsy precision oncotherapy to third-line systemic therapy with drug regimes selected partly by tissue biopsy precision oncotherapy, in a retrospective real-life study of 106 unresectable CRCLM patients. Drug regimens for HAI/target therapy were selected by assessing the sensitivity of purified circulating tumor cell (CTCs) to 5-fluorouracil, carboplatin, cisplatin, oxaliplatin, irinotecan, doxorubicin, mitomycin, raltitrexed, and melphalan in-vitro and by real-time qRT-PCR gene expression assays, and for the Systemic therapy cohort were selected by age, comorbidity, performance status, and absence of RAS mutations. Therapeutic responses, adverse events, and quality of life were evaluated by RECIST 1.1, CTCAE 4.03, and ECOG criteria, respectively, and chemo-filtration performed following HAI to reduce systemic toxic effects. HAI/target therapy with drugs selected by liquid biopsy precision oncotherapy (44 patients), resulted in 2.27% CRs, 38.63% PRs, 56.81% SD,s and 2.27% PDs; ECOG 2 to 1 improvement, but no infusion-related technical or vascular complications, or deaths. Systemic therapy (62 patients) resulted in 1.6% CRs, 17.74% PRs, 37.09% SDs, and 45.16% PDs; more grade 1-2 adverse events and 4.84% ECOG 1 to 2 worsening. The median 5 month PFS in the HAI/target therapy cohort was significantly longer than 3 months in the systemic cohort (P < 0.007) and the median 14 month survival in the HAI/target therapy cohort was longer than 8.5 months in the systemic therapy cohort but not statistically significant. Multivariate analysis identified ECOG grade 2 as the most unfavourable survival prognostic factor in both cohorts. HAI plus chemo-filtration followed by target therapy, with drug regimens selected by liquid biopsy precision oncotherapy, is a safe and efficacious alternative therapeutic strategy for unresectable CRCLM in progression after two lines of systemic therapy and should be considered for a multicentre prospective phase III study, to fully confirm this potential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Melphalan, powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder