Merck
Metformin enhances the sensitivity of colorectal cancer cells to cisplatin through ROS-mediated PI3K/Akt signaling pathway.
Gene (2020-03-31)
Pei Zhang, Surong Zhao, Xingyue Lu, Zongfen Shi, Hao Liu, Bing Zhu
ABSTRAKT
Metformin and cisplatin have been widely studied as antitumor agents. However, the effect of metformin combined with cisplatin has not been investigated in colorectal cancer (CRC) cells. This study was aimed to explore the effect of metformin or/and cisplatin on cell viability, apoptosis, and the related signaling pathways in CRC SW480 and SW620 cells. We found that metformin or cisplatin inhibited cell viability of SW480 and SW620 cells in a concentration- and time-dependent manner. Furthermore, metformin combined with cisplatin obviously inhibited cell viability, decreased colony formation, induced apoptosis, mediated cleavage of caspase-9, caspase-3 and PARP, activated mitochondrial membrane potential, downregulated Mcl-1 and Bcl-2 expression, upregulated Bak and Bax expression, and increased reactive oxygen species (ROS) production, compared to the individual agent in SW480 and SW620 cells, which were attenuated by N-acetyl-L-cysteine (NAC), a ROS scavenger. Moreover, NAC could recover the downregulation of p-PI3K and p-Akt treated with combination of metformin and cisplatin, which subsequently activated the PI3K/Akt signaling pathway. Taken together, our results demonstrated that metformin enhanced the sensitivity of CRC cells to cisplatin through ROS-mediated PI3K/Akt signaling pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
JC-1, powder or solid (Crystals)