Merck
The effect and mechanism of 19S proteasome PSMD11/Rpn6 subunit in D-Galactose induced mimetic aging models.
Experimental cell research (2020-05-26)
Han Wu, Haiying Sun, Zuhong He, Xi Chen, Yongqin Li, Xueyan Zhao, Wen Kong, Weijia Kong
ABSTRAKT
Regulating proteasome activity is a potent therapeutic aspect of age-related hearing loss, which has been proven to protect neurons from age-related damaging. PSMD11, subunit of the 19S proteasome regulatory particle, is known to mainly up-regulate proteasome activity and prolong aging. However, the mechanism of PSMD11 in age-related hearing loss has not been deeply explored. In the present study, we explore the function and mechanism of PSMD11 protecting neurons in d-Galactose (D-Gal) mimetic aging models. Age-related pathologies were detected by Taq-PCR, ABR, Transmission electron microscopy, toluidine blue and β-galactosidase staining. The relative expressions of the proteins were explored by Western blotting, oxyblot, immunoprecipitation and immunofluorescence. Flow cytometry was used to manifest the oxidative state. We discovered that proteasome activity was impaired with aging, and that ROS and toxic protein accumulated in D-Gal induced aging models. PSMD11 changed with aging, and was associated with the metabolism of proteasome activity in the D-Gal treated models. Moreover, the knockdown or overexpression of PSMD11 was sufficient to change the oxidative state caused by D-Gal. Our results also demonstrated that PSMD11 could bond to AMPKα1/2 in the auditory cortex and PC12 cells, and AMPKα2 but not AMPKα1 was efficient to regulate the function of PSMD11. Deeper insights into the mechanisms of regulating PSMD11 for the anti-aging process are needed, and may offer novel therapeutic methods for central presbycusis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder