Merck
Lipocalin-type prostaglandin D synthase regulates light-induced phase advance of the central circadian rhythm in mice.
Communications biology (2020-10-10)
Chihiro Kawaguchi, Norihito Shintani, Atsuko Hayata-Takano, Michiyoshi Hatanaka, Ai Kuromi, Reiko Nakamura, Yui Yamano, Yusuke Shintani, Katsuya Nagai, Soken Tsuchiya, Yukihiko Sugimoto, Atsushi Ichikawa, Yasushi Okuno, Yoshihiro Urade, Hiroyuki Hirai, Kin-Ya Nagata, Masataka Nakamura, Shuh Narumiya, Takanobu Nakazawa, Atsushi Kasai, Yukio Ago, Kazuhiro Takuma, Akemichi Baba, Hitoshi Hashimoto
ABSTRAKT
We previously showed that mice lacking pituitary adenylate cyclase-activating polypeptide (PACAP) exhibit attenuated light-induced phase shift. To explore the underlying mechanisms, we performed gene expression analysis of laser capture microdissected suprachiasmatic nuclei (SCNs) and found that lipocalin-type prostaglandin (PG) D synthase (L-PGDS) is involved in the impaired response to light stimulation in the late subjective night in PACAP-deficient mice. L-PGDS-deficient mice also showed impaired light-induced phase advance, but normal phase delay and nonvisual light responses. Then, we examined the receptors involved in the response and observed that mice deficient for type 2 PGD2 receptor DP2/CRTH2 (chemoattractant receptor homologous molecule expressed on Th2 cells) show impaired light-induced phase advance. Concordant results were observed using the selective DP2/CRTH2 antagonist CAY10471. These results indicate that L-PGDS is involved in a mechanism of light-induced phase advance via DP2/CRTH2 signaling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BW A868C, ≥98% (HPLC)