Merck
  • Chemo-photodynamic therapy by pulmonary delivery of gefitinib nanoparticles and 5-aminolevulinic acid for treatment of primary lung cancer of rats.
Chemo-photodynamic therapy by pulmonary delivery of gefitinib nanoparticles and 5-aminolevulinic acid for treatment of primary lung cancer of rats.
Photodiagnosis and photodynamic therapy (2020-05-15)
Tongtong Zhang, Jianwei Bao, Mengmeng Zhang, Yuanyuan Ge, Jianming Wei, Yu Li, Weixia Wang, Miao Li, Yiguang Jin
ABSTRAKT
Lung cancer is a severe disease with high mortality. Chemotherapy is one major treatment for lung cancer. However, systemic chemotherapeutics usually distribute throughout the body without specific lung distribution so that serious side effects are unavoidable. Photodynamic therapy (PDT) is occasionally used for lung cancer treatment but photosensitizers are also systemically administered and the bronchoscopic intervention under anesthesia may hurt lung tissues. Here, we combined inhaled chemotherapeutics and photosensitizers for chemo-photodynamic therapy (CPDT) of primary lung cancer of rats with external laser light irradiation. Gefitinib PLGA nanoparticles (GNPs) were prepared. The anti-cancer effects of GNPs and/or a common photosensitizer 5-aminolevulinic acid (5-ALA) were explored on A549 cells (adenocarcinomic human alveolar basal epithelial cells) and primary lung cancer rats after intratracheal administration. External light irradiation was applied due to its higher safety compared to internal light irradiation that may result in injuries after a laser optic fiber was intubated into the lung. The remarkable synergistic effect of CPDT was confirmed although the single therapies were also effective, where the high anti-lung cancer effects were shown and some typical lung cancer markers, including CD31, VEGF, NF-κB p65 and Bcl-2, significantly decreased. Moreover, the treatments attenuated inflammation with the downregulation of TNF-α. The combination of pulmonary drug delivery and chemo-photodynamic therapy is a promising strategy for treatment of lung cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder