Merck
Electrical stimulation of the lateral cerebellar nucleus promotes neurogenesis in rats after motor cortical ischemia.
Scientific reports (2020-10-08)
Zheng Wu, Fangling Sun, Zijie Li, Min Liu, Xin Tian, Deyu Guo, Penghu Wei, Yongzhi Shan, Tingting Liu, Min Guo, Zixin Zhu, Wenrong Zheng, Yufeng Wang, Guoguang Zhao, Wen Wang
ABSTRAKT
Deep brain stimulation (DBS) has been tentatively explored to promote motor recovery after stroke. Stroke could transiently activate endogenous self-repair processes, including neurogenesis in the subventricular zone (SVZ). In this regard, it is of considerable clinical interest to study whether DBS of the lateral cerebellar nucleus (LCN) could promote neurogenesis in the SVZ for functional recovery after stroke. In the present study, rats were trained on the pasta matrix reaching task and the ladder rung walking task before surgery. And then an electrode was implanted in the LCN following cortical ischemia induced by endothelin-1 injection. After 1 week of recovery, LCN DBS coupled with motor training for two weeks promoted motor function recovery, and reduced the infarct volumes post-ischemia. LCN DBS augmented poststroke neurogenetic responses, characterized by proliferation of neural progenitor cells (NPCs) and neuroblasts in the SVZ and subsequent differentiation into neurons in the ischemic penumbra at 21 days poststroke. DBS with the same stimulus parameters at 1 month after ischemia could also increase nascent neuroblasts in the SVZ and newly matured neurons in the perilesional cortex at 42 days poststroke. These results suggest that LCN DBS promotes endogenous neurogenesis for neurorestoration after cortical ischemia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Roche
Anty-bromodeoksyurydyna, from mouse IgG1 (clone: BMC9318)