Merck
Artesunate reverses LPS tolerance by promoting ULK1-mediated autophagy through interference with the CaMKII-IP3R-CaMKKβ pathway.
International immunopharmacology (2020-08-08)
Xin Liu, Jiaqi Wu, Ning Wang, Lin Xia, Shijun Fan, Yongling Lu, Xiaoli Chen, Shenglan Shang, Yongjun Yang, Qianying Huang, Qian Chen, Hong Zhou, Jiang Zheng
ABSTRAKT
The progress of sepsis is increasingly recognized by the transition from early hyperinflammation to long term immunosuppression, which is characterized in innate immune cells by diminished responsiveness termed as lipopolysaccharide (LPS) tolerance. In this study, we investigated the ability of the antimalarial drug artesunate to reverse LPS tolerance and explored the underlying mechanisms. Initially, we detected a dramatic decline in autophagy accompanied by decreased cytokine production and impaired bacterial clearance by LPS tolerant macrophages. Then we demonstrated that artesunate restored cytokine production and enhanced bacterial clearance by inducing autophagy. Moreover, artesunate caused greater suppression of inhibitory phosphorylation than of activating phosphorylation of Unc-51 like autophagy activating kinase 1 (ULK1), a kinase that is essential for initiating autophagy through the inhibition of excessive AMP-activated protein kinase (AMPK) activation. This effect was shown to be achieved by suppression of Ca2+/calmodulin-dependent protein kinase II (CaMKII) phosphorylation, resulting in reduction of the inositol 1,4,5-triphate receptor (IP3R) dependent Ca2+ release from the endoplasmic reticulum (ER) and inhibiting the overactive CaMKKβ-AMPK cascade. Administration of artesunate also upregulated autophagy and reversed the tolerant status in LPS tolerant mice. In summary, our findings reveal a novel immunopharmacological action of artesunate to reverse LPS tolerance by restoring autophagy. Our results may also indicate the significance of autophagy induction for treating immunosuppression in sepsis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Supelco
Ponceau 6R, analytical standard