Merck
RBMX is required for activation of ATR on repetitive DNAs to maintain genome stability.
Cell death and differentiation (2020-06-05)
Tian Zheng, Haoxian Zhou, Xiaocui Li, Di Peng, Yiding Yang, Yanru Zeng, Haiying Liu, Jian Ren, Yong Zhao
ABSTRAKT
ATR is a master regulator of cell response to replication stress. Adequate activation of ATR is essential for preventing genome aberrance induced by replication defect. However, the mechanism underlying ATR activation is not fully understood. Here, we identify that RBMX is an ssDNA binding protein that orchestrates a novel pathway to activate ATR. Using super-resolution STORM, we observe that RBMX and RPA bind to adjacent but nonoverlapping sites on ssDNA in response to replication stress. RBMX then binds to and facilitates positioning of TopBP1, which activates nearby ATR associated with RPA. In addition, ATR activation by ssDNA-RBMX-TopBP1 is independent of ssDNA-dsDNA junction and 9-1-1 complex. ChIP-seq analysis reveals that RBMX/RPA are highly enriched on repetitive DNAs, which are considered as fragile sites with high replication stress. RBMX depletion leads to defective localization of TopBP1 to replication stressed sites and inadequate activation of ATR. Furthermore, cells with deficient RBMX demonstrate replication defect, leading to formation of micronuclei and a high rate of sister-chromatin exchange, indicative of genome instability. Together, the results identify a new ssDNA-RBMX-TopBP1 pathway that is specifically required for activation of ATR on repetitive DNAs. Therefore, RBMX is a key factor to ensure genome stability during replication.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Replication Protein A Antibody, clone RPA34-20, clone RPA34-20, from mouse
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)