- 2-(Arylmethyl)-3-substituted quinuclidines as selective alpha 7 nicotinic receptor ligands.
2-(Arylmethyl)-3-substituted quinuclidines as selective alpha 7 nicotinic receptor ligands.
Bioorganic & medicinal chemistry letters (2005-04-06)
Anatoly Mazurov, Jozef Klucik, Lan Miao, Teresa Y Phillips, Angela Seamans, Jeffrey D Schmitt, Terry A Hauser, Raymond T Johnson, Craig Miller
PMID15808471
ABSTRAKT
A series of 2-(arylmethyl)-3-substituted quinuclidines was developed as alpha7 neuronal nicotinic acetylcholine receptor (nAChR) agonists based on a putative pharmacophore model. The series is highly selective for the alpha7 over other nAChRs (e.g., the alpha4beta2 of the CNS, and the muscle and ganglionic subtypes) and is functionally tunable at alpha7. One member of the series, (+)-N-(1-azabicyclo[2.2.2]oct-3-yl)benzo[b]furan-2-carboxamide (+)-8l), has potent agonistic activity for the alpha7 nAChR (EC(50)=33nM, I(max)=1.0), at concentrations below those that result in desensitization.