Merck
Synthesis and in vitro study of novel Mannich bases as antibacterial agents.
Bioorganic & medicinal chemistry letters (2004-12-08)
Sheela Joshi, Navita Khosla, Deepak Khare, Rakesh Sharda
ABSTRAKT
A series of novel Mannich bases derived from 5-chloro-2-methoxybenzamide and sulfonamides/amines have been synthesised and the antibacterial activities were evaluated against various Gram positive and Gram negative strains of bacteria. Some of the synthesized compounds showed superior in vitro activities as compared to their parent sulfonamides.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%