Merck
  • Design, synthesis, and biological evaluation of the N-diarylalkenyl-piperidinecarboxylic acid derivatives as GABA uptake inhibitors (I).
Design, synthesis, and biological evaluation of the N-diarylalkenyl-piperidinecarboxylic acid derivatives as GABA uptake inhibitors (I).
Bioorganic & medicinal chemistry letters (2005-10-26)
Jianbin Zheng, Ren Wen, Xiaomin Luo, Guoqiang Lin, Jiange Zhang, Linfeng Xu, Lihe Guo, Hualiang Jiang
ABSTRAKT
Twenty novel N-diarylalkenyl-piperidinecarboxylic acid derivatives were synthesized and evaluated as gamma-aminobutyric acid uptake inhibitors. The biological assay showed that (R)-1-[4,4-bis(3-phenoxymethyl-2-thienyl)-3-butenyl]-3-piperidinecarboxylic hydrochloride possessed almost as strong GAT1 inhibitory activity as tiagabine. The synthesis and structure-activity relationships are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(R)-(–)-3-Piperidinecarboxylic acid, 97%