Merck
Possible involvement of hypothalamic nucleobindin-2 in hyperphagic feeding in Tsumura Suzuki obese diabetes mice.
Biological & pharmaceutical bulletin (2012-10-06)
Shigeo Miyata, Nao Yamada, Tomie Kawada
ABSTRAKT
The aim of this study was to clarify the hypothalamic neuropeptides that are associated with hyperphagic feeding in Tsumura Suzuki Obese Diabetes (TSOD) mice, a model of type 2 diabetes with polygenic abnormalities. TSOD mice showed an increase in body weight and hyperleptinemia from 1 month of age and hyperphagic feeding, hyperglycemia, hyperlipidemia and hyperinsulinemia from 3 to 12 months of age compared with age-matched non-diabetic control Tsumura Suzuki Non Obesity (TSNO) mice. The mRNA level of nucleobindin-2 (NUCB2), the precursor of the anorexigenic neuropeptide nesfatin-1, was significantly decreased in the hypothalamus of TSOD mice compared with that in TSNO mice from 3 to 12 months of age. The protein level of NUCB2 was significantly decreased in the hypothalamus of TSOD mice compared with that in TSNO mice at 3 months of age. The mRNA levels of galanin, melanin-concentrating hormone, neuropeptide Y, and pro-opiomelanocortin were significantly changed in the hypothalamus in TSOD mice at several time points. Another model of type 2 diabetes, db/db mice, which is a mutant mouse that lacks a functional leptin receptor, showed hyperphagic feeding but no change in hypothalamic NUCB2 mRNA compared with non-diabetic control db/+ mice. The results suggest that the disrupted control of hypothalamic NUCB2-mediated signaling may contribute to hyperphagic feeding in TSOD mice. In addition, the mechanism for the development of hyperphagic feeding in TSOD mice is different than that in db/db mice.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution