Merck
  • Design and synthesis of novel quinolinone-3-aminoamides and their alpha-lipoic acid adducts as antioxidant and anti-inflammatory agents.
Design and synthesis of novel quinolinone-3-aminoamides and their alpha-lipoic acid adducts as antioxidant and anti-inflammatory agents.
Journal of medicinal chemistry (2007-04-21)
Anastasia Detsi, Dionysia Bouloumbasi, Kyriakos C Prousis, Maria Koufaki, Giorgos Athanasellis, Georgia Melagraki, Antreas Afantitis, Olga Igglessi-Markopoulou, Christos Kontogiorgis, Dimitra J Hadjipavlou-Litina
ABSTRAKT
A series of N-substituted-quinolinone-3-aminoamides and their hybrids containing the alpha-lipoic acid functionality were designed and synthesized as potential bifunctional agents combining antioxidant and anti-inflammatory activity. The new compounds were evaluated for their antioxidant activity and for their ability to inhibit in vitro lipoxygenase as well as for their anti-inflammatory activity in vivo. In general, the derivatives were found to be potent antioxidant or anti-inflammatory agents. The results are discussed in terms of structure-activity relationships and an attempt is made to define the structural features required for activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Supelco
Kwas(R)-(+)-α-liponowy, analytical standard