Merck
CD163 expression defines specific, IRF8-dependent, immune-modulatory macrophages in the bone marrow.
The Journal of allergy and clinical immunology (2020-03-23)
Lena Fischer-Riepe, Niklas Daber, Jonas Schulte-Schrepping, Bruna Caroline Véras De Carvalho, Antonella Russo, Michele Pohlen, Josephine Fischer, Achmet Imam Chasan, Marc Wolf, Thomas Ulas, Shirin Glander, Christian Schulz, Boris Skryabin, Andreas Wollbrink Dipl-Ing, Nadine Steingraeber, Christopher Stremmel, Megan Koehle, Florian Gärtner, Sabine Vettorazzi, Dirk Holzinger, Joachim Gross, Frank Rosenbauer, Monika Stoll, Silke Niemann, Jan Tuckermann, Joachim L Schultze, Johannes Roth, Katarzyna Barczyk-Kahlert
ABSTRAKT
Scavenger receptor CD163 is exclusively expressed on monocytes/macrophages and is widely used as a marker for alternatively activated macrophages. However, the role of CD163 is not yet clear. We sought to examine the function of CD163 in steady-state as well as in sterile and infectious inflammation. Expression of CD163 was analyzed under normal and inflammatory conditions in mice. Functional relevance of CD163 was investigated in models of inflammation in wild-type and CD163-/- mice. We describe a subpopulation of bone marrow-resident macrophages (BMRMs) characterized by a high expression of CD163 and functionally distinct from classical bone marrow-derived macrophages. Development of CD163+ BMRMs is strictly dependent on IFN regulatory factor-8. CD163+ BMRMs show a specific transcriptome and cytokine secretion pattern demonstrating a specific immunomodulatory profile of these cells. Accordingly, CD163-/- mice show a stronger inflammation in allergic contact dermatitis, indicating a regulatory role of CD163. However, CD163-/- mice are highly susceptible to S aureus infections, demonstrating the relevance of CD163 for antimicrobial defense as well. Our data indicate that anti-inflammatory and immunosuppressive mechanisms are not necessarily associated with a decreased antimicrobial activity. In contrast, our data define a novel macrophage population that controls overwhelming inflammation on one hand but is also necessary for an effective control of infections on the other hand.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%