Merck
Synthesis, antifungal and antimicrobial activity of alkylphospholipids.
Bioorganic & medicinal chemistry (2007-05-29)
Daniel Obando, Fred Widmer, Lesley C Wright, Tania C Sorrell, Katrina A Jolliffe
ABSTRAKT
The antifungal, antibacterial and haemolytic activity of a series of alkylphosphocholines (e.g., miltefosine) and alkylglycerophosphocholines (e.g., edelfosine) has been investigated. These compound classes exhibit significant antifungal and moderate antibacterial activities. Several new alkylphosphocholine derivatives with amide or ester bonds in the alkyl chain have been synthesised. These compounds show much lower haemolytic activity than miltefosine. Alkylphosphocholines and alkylglycerophosphocholines show significant promise as novel orally available antifungal and antibacterial therapeutics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Miltefosine, ≥98% (perchloric acid titration)