Merck
  • Prevalence of plasmid-mediated quinolone resistance determinants QnrA, QnrB, and QnrS among clinical isolates of Enterobacter cloacae in a Taiwanese hospital.
Prevalence of plasmid-mediated quinolone resistance determinants QnrA, QnrB, and QnrS among clinical isolates of Enterobacter cloacae in a Taiwanese hospital.
Antimicrobial agents and chemotherapy (2007-01-24)
Jiunn-Jong Wu, Wen-Chien Ko, Shu-Huei Tsai, Jing-Jou Yan
ABSTRAKT
The prevalence of three plasmid-mediated quinolone resistance determinants, QnrA, QnrB, and QnrS, among 526 nonreplicate clinical isolates of Enterobacter cloacae collected at a Taiwanese university hospital in 2004 was determined by PCR and colony hybridization, and the association of Qnr with the IMP-8 metallo-beta-lactamase was investigated. Eighty-six (16.3%) of all isolates were qnr positive, and the qnrA1-like, qnrB2-like, and qnrS1-like genes were detected alone or in combination in 3 (0.6%), 53 (10.1%), and 34 (6.5%) isolates, respectively. Among 149 putative extended-spectrum-beta-lactamase-producing isolates, 59 (39.6%) isolates, all of which were SHV-12 producers, harbored qnrA (0.7%; 1 isolate), qnrB (28.9%; 43 isolates), or qnrS (12.1%; 18 isolates). Forty-four (78.6%) of 56 IMP-8 producers carried qnrB (58.9%; 33 isolates), qnrS (25.0%; 14 isolates), or both. PCR and sequence analysis revealed that qnrA1 was located in a complex sul1-type integron that contains dhr15, aadA2, qacEDelta1, sul1, orf513, qnrA1, ampR, and qacEDelta1. Conjugation experiments revealed the coexistence of qnrB and bla(IMP-8) on the transferred plasmids and the absence of beta-lactamase content on the transferred qnrS-positive plasmids. The transferred bla(IMP-8)-positive plasmids with and without qnrB had very similar restriction patterns, suggesting the horizontal mobility of qnrB. Pulsed-field gel electrophoresis showed six major patterns among the 44 qnr-positive IMP-8-producing isolates. Thus, the extremely high prevalence of qnr among the metallo-beta-lactamase-producing E. cloacae isolates in the hospital may be due mainly to the intrahospital spread of a few clones and the dissemination of plasmids containing both qnrB and blaIMP-8.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Nalidixic acid, ≥98%
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)