Merck
  • Developing a biodegradable tricalcium silicate/glucono-delta-lactone/calcium sulfate dihydrate composite cement with high preliminary mechanical property for bone filling.
Developing a biodegradable tricalcium silicate/glucono-delta-lactone/calcium sulfate dihydrate composite cement with high preliminary mechanical property for bone filling.
Materials science & engineering. C, Materials for biological applications (2020-12-17)
Zhengwen Ding, Wenjing Xi, Mizhi Ji, Hong Chen, Qiyi Zhang, Yonggang Yan
ABSTRAKT
Bone cements with the feature of easily shaping could ideally match the defect site and prevent the ingrowth of fibrous tissue. In this manuscript, a biodegradable tricalcium silicate (C3S)/glucono-delta-lactone (GDL)/calcium sulfate dihydrate (CSD) organic-inorganic composite cement was fabricated with shorter setting time (less than 15 min) and high preliminary mechanical property (5.27 MPa in the first hour). Many methods were applied to study the physicochemical and biological properties of the cement in vitro. The weight loss in PBS can reach 58% after 12 weeks soaking indicating the better biodegradability. The excellent bioactivity in vitro was emerging after the cement was soaked in the simulated body fluid. The cell experiments showed that suitable concentration of the extract liquid of cement was conducive to the proliferation, differentiation and extracellular matrix calcification of the mouse bone marrow stromal cells. Briefly, the C3S/GDL/CSD composite cement would have the bright capacity for bone filling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder