Merck
Downregulation of lncRNA ZFAS1 inhibits the hallmarks of thyroid carcinoma via the regulation of miR‑302‑3p on cyclin D1.
Molecular medicine reports (2020-11-13)
Wenjing Chen, Lili Zhai, Huiming Liu, Yuting Li, Qi Zhang, Dandan Xu, Weiye Fan
ABSTRAKT
At present, treatment options for thyroid carcinoma remain limited. The present study aimed to investigate the role of ZFAS1 in various major hallmarks of cancer and the underlying mechanisms in thyroid carcinoma cells. The interactions between long non‑coding RNAs (lncRNAs), microRNAs (miRs) and target genes were predicted by bioinformatics and confirmed by performing dual‑luciferase assays. The mRNA and protein expressions were determined by reverse transcription‑quantitative PCR and western blotting. Cell invasion, migration, and viability were evaluated via Transwell, wound‑healing and Cell Counting Kit‑8 assays, respectively. The results demonstrated that lncRNA ZFAS1 expression was upregulated in thyroid carcinoma tissues, TT and SW579 cells, and was associated with the proliferation of these two cell lines. Notably, downregulation ZFAS1 reduced migration and invasion, and reversed the promotive effects of miR‑302a‑3p inhibitor on the proliferation, migration and invasion of TT and SW579 cells. Moreover, cyclin D1 (CCND1) is targeted by miR‑302a‑3p, and was regulated by ZFAS1. In addition, the downregulation of ZFAS1 not only reversed the promotive effects of miR‑302a‑3p inhibitor on CCND1 expression and the epithelial‑mesenchymal transition (EMT) of TT and SW579 cells, but also targeted and increased the expression of miR‑302a‑3p, and further reduced the expression of CCND1, resulting in suppression of the proliferation, migration, invasion and EMT of thyroid carcinoma cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder